1-(4-Fluorophenyl)-2-imidazolidinone

Supplier: APOLLO SCIENTIFIC
Warning

PC8783-1G PC8783-250MG
76561-190EA 51 USD
76561-190 76561-056
1-(4-Fluorophenyl)-2-imidazolidinone
1-(4-Fluorophenyl)-2-imidazolidinone
Formula: C₉H₉FN₂O
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD03787796
CAS Number: 53159-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR