2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(2-Cyano-3-fluoro)-2-(thien-2-yl)acetonitrile

PC8446-1G PC8446-100MG
76563-274EA 177 USD
76563-274 76559-862
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile 97%
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile
Formula: C₁₃H₇FN₂S
MW: 242.28 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00231908
CAS Number: 306976-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR