3-(Trifluoromethyl)benzamidoxime 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: N'-Hydroxy-3-(trifluoromethyl)-benzenecarboximidamide

PC7507L-100G PC7507L-5G PC7507L-25G
76562-756EA 162 USD
76562-756 76563-006 76560-594
3-(Trifluoromethyl)benzamidoxime 97%
3-(Trifluoromethyl)benzamidoxime
Formula: C₈H₇F₃N₂O
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00068189
CAS Number: 40067-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR