2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

PC7181-5G PC7181-1G
76561-688EA 13.33 USD
76561-688 76561-960
2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD07784336
CAS Number: 157834-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR