3-Bromo-5-fluoro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

PC5508-250MG
76560-150EA 54 USD
76560-150
3-Bromo-5-fluoro-1H-indazole
3-Bromo-5-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781639
CAS Number: 885519-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR