2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC

Warning

PC4034-1G PC4034-500MG
76562-884EA 171 USD
76562-884 76560-930
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole
2-Hydrazino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formula: C₃H₃F₃N₄S
Storage Temperature: Refrigerator
MDL Number: MFCD07786303
CAS Number: 299440-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR