2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC

PC303035-1G PC303035-10G PC303035-5G
76562-444EA 171 USD
76562-444 76562-138 76562-382
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile
Formula: C₉H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD22201044
CAS Number: 1373920-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR