4-Ethoxy-2-fluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC302987-1G PC302987-10G PC302987-5G
76560-634EA 51 USD
76560-634 76561-606 76559-914
4-Ethoxy-2-fluorobenzonitrile 98%
4-Ethoxy-2-fluorobenzonitrile
Formula: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD13196591
CAS Number: 186191-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR