4-Ethoxy-3-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC302335-5G PC302335-1G PC302335-10G
76560-182EA 124 USD
76560-182 76562-718 76560-230
4-Ethoxy-3-(trifluoromethyl)benzonitrile 98%
4-Ethoxy-3-(trifluoromethyl)benzonitrile
Formula: C₁₀H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD13196579
CAS Number: 1206593-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR