2,3-Dichloro-6-(trifluoromethyl)aniline 98%

Supplier: APOLLO SCIENTIFIC

PC302220-1G PC302220-10G PC302220-5G
76562-564EA 114 USD
76562-564 76560-244 76561-474
2,3-Dichloro-6-(trifluoromethyl)aniline 98%
2,3-Dichloro-6-(trifluoromethyl)aniline
Formula: C₇H₄Cl₂F₃N
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD09832277
CAS Number: 1017777-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR