2-Fluoro-5-methylanisole 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Fluoro-2-methoxy-4-methylbenzene4-Fluoro-3-methoxytoluene

PC0083-1G
76561-628EA 13 USD
76561-628
2-Fluoro-5-methylanisole 98%
2-Fluoro-5-methylanisole
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 93…95 °C/30mmHg
Density: 1.07 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00665791
CAS Number: 63762-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR