4-Amino-3-chlorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR9762-5G OR9762-100G OR9762-25G
76559-798EA 13.33 USD
76559-798 76561-318 76561-828
4-Amino-3-chlorobenzonitrile
4-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00052917
CAS Number: 21803-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR