N-Methyl-3-nitrobenzenesulfonamide 98%

Supplier: APOLLO SCIENTIFIC

OR5808-1G OR5808-5G
76560-500EA 63 USD
76560-500 76562-604
N-Methyl-3-nitrobenzenesulfonamide 98%
N-Methyl-3-nitrobenzenesulphonamide
Formula: C₇H₈N₂O₄S
Melting Pt: 110…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00474673
CAS Number: 58955-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR