Isoquinoline-1-carbonitrile 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Isoquinolinecarbonitrile

OR5801-1G
76559-454EA 30 USD
76559-454
Isoquinoline-1-carbonitrile 98%
Isoquinoline-1-carbonitrile
Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00134166
CAS Number: 1198-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR