N,N-Dimethyl 4-boronobenzenesulfonamide 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(N,N-Dimethylsulfamoyl)phenylboronic acid4-(Dimethylsulfamoyl)benzeneboronic acid

OR3969-250MG
76560-322EA 31 USD
76560-322
N,N-Dimethyl 4-boronobenzenesulfonamide 95%
N,N-Dimethyl 4-boronobenzenesulphonamide
Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD06659849
CAS Number: 486422-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR