2-(Cyanomethoxy)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR361023-100MG
76560-790EA 114 USD
76560-790
2-(Cyanomethoxy)phenylboronic acid 98%
2-(Cyanomethoxy)phenylboronic acid
Formula: C₈H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09908171
CAS Number: 947533-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR