4-Chlorosalicylaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Chloro-2-hydroxybenzaldehyde

OR30987-1G OR30987-5G
76575-412EA 16 USD
76575-412 76575-414
4-Chlorosalicylaldehyde
4-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.5664 g/mol
Boiling Pt: 230 °C
Melting Pt: 47…52 °C
Storage Temperature: Ambient
MDL Number: MFCD06252499
CAS Number: 2420-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR