3-Amino-4-chloro-1-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

OR30983-1G
76575-406EA 92 USD
76575-406
3-Amino-4-chloro-1-methyl-1H-indazole
3-Amino-4-chloro-1-methyl-1H-indazole
Formula: C₈H₈ClN₃
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09864571
CAS Number: 959240-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR