N-Benzyl-N-(3-butenyl)amine 95%

Supplier: APOLLO SCIENTIFIC

Warning

OR30936-250MG OR30936-1G
76575-346EA 113 USD
76575-346 76575-348
N-Benzyl-N-(3-butenyl)amine 95%
N-Benzyl-N-(3-butenyl)amine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 65 °C/0.75mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD01249643
CAS Number: 17150-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR