1H-Indazole-7-boronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

OR308056-250MG OR308056-1G
76575-134EA 194 USD
76575-134 76575-132
1H-Indazole-7-boronic acid pinacol ester 97%
1H-Indazole-7-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD08059264
CAS Number: 915411-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR