5-Bromo-6-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

OR30742-5G OR30742-1G OR30742-25G
76575-016EA 25 USD
76575-016 76575-018 76575-020
5-Bromo-6-methyl-1H-indazole
5-Bromo-6-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD09026991
CAS Number: 885223-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR