1H-Indazole-6-boronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

OR30736-500MG OR30736-1G
76575-002EA 37 USD
76575-002 76575-004
1H-Indazole-6-boronic acid pinacol ester 97%
1H-Indazole-6-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
Melting Pt: 134…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368067
CAS Number: 937049-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR