4-Cyclopropylphenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (4-Cyclopropylphenyl)boronic acid

OR307250-1G OR307250-250MG
76574-878EA 87 USD
76574-878 76574-880
4-Cyclopropylphenylboronic acid
4-Cyclopropylphenylboronic acid
Formula: C₉H₁₁BO₂
MW: 161.9934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01309064
CAS Number: 302333-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR