4-Bromo-3-nitrotoluene 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Bromo-4-methyl-2-nitrobenzene

OR3068-100G OR3068-25G
76574-686EA 63 USD
76574-686 76574-684
4-Bromo-3-nitrotoluene 99%
4-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 152 °C/14mmHg
Melting Pt: 31…33 °C
Density: 1.578 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR