Octachloronaphthalene 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR30671-5G OR30671-1G OR30671-25G
76574-676EA 25 USD
76574-676 76574-678 76574-680
Octachloronaphthalene 98%
Octachloronaphthalene
Formula: C₁₀Cl₈
MW: 403.73 g/mol
Boiling Pt: 440 °C
Melting Pt: 194…197 °C
Density: 2 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
CAS Number: 2234-13-1
EINECS: 218-778-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR