4-(1-Naphthyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-(1-Naphthyl)benzeneboronic acid

OR30657-100G OR30657-5G OR30657-25G
76574-606EA 103 USD
76574-606 76574-604 76574-608
4-(1-Naphthyl)phenylboronic acid
4-(1-Naphthyl)phenylboronic acid
Formula: C₁₆H₁₃BO₂
MW: 248.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669639
CAS Number: 870774-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR