5-Bromoimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR30602-1G OR30602-5G
76574-562EA 77 USD
76574-562 76574-560
5-Bromoimidazo[1,2-a]pyridine
5-Bromoimidazo[1,2-a]pyridine
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD08275698
CAS Number: 69214-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR