1-Methyl-1H-indazole-7-boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (1-Methyl-1H-indazol-7-yl)boronic acid

OR30588-1G OR30588-250MG
76574-548EA 126 USD
76574-548 76574-546
1-Methyl-1H-indazole-7-boronic acid
1-Methyl-1H-indazole-7-boronic acid
Formula: C₈H₉BN₂O₂
MW: 175.9803 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09870054
CAS Number: 1001907-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR