4-Amino-2-chloro-5-methylpyridine 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Chloro-5-methylpyridin-4-amine

OR305585-25G OR305585-5G OR305585-1G
76574-500EA 224 USD
76574-500 76574-498 76574-496
4-Amino-2-chloro-5-methylpyridine 98%
4-Amino-2-chloro-5-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.5862 g/mol
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD11977435
CAS Number: 79055-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR