2-(Chloromethyl)quinoline hydrochloride

Supplier: APOLLO SCIENTIFIC

Danger

OR30556-1G OR30556-5G OR30556-25G
76574-478EA 37 USD
76574-478 76574-476 76574-474
2-(Chloromethyl)quinoline hydrochloride
2-(Chloromethyl)quinoline hydrochloride
Formula: C₁₀H₉Cl₂N
MW: 214.10 g/mol
Melting Pt: 183…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012734
CAS Number: 3747-74-8
EINECS: 223-145-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR