(±)-3-Methoxy-1,2-propanediol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (±)-3-Methoxy-1,2-dihydroxy propane

OR30517-5G OR30517-25G
76574-378EA 58 USD
76574-378 76574-380
(±)-3-Methoxy-1,2-propanediol 98%
(±)-3-Methoxy-1,2-propanediol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 218…220 °C
Density: 1.114 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004719
CAS Number: 623-39-2
EINECS: 210-791-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR