4-Methoxy-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

OR305095-1G
76574-350EA 83 USD
76574-350
4-Methoxy-1H-indazole
4-Methoxy-1H-indazole
Formula: C₈H₈N₂O
MW: 148.1619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261131
CAS Number: 351210-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR