3-Iodo-6-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Iodo-6-nitroindazole

OR305075-5G OR305075-1G
76574-338EA 88 USD
76574-338 76574-336
3-Iodo-6-nitro-1H-indazole
3-Iodo-6-nitro-1H-indazole
Formula: C₇H₄IN₃O₂
MW: 289.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781652
CAS Number: 70315-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR