2-Indanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Hydroxyindane

OR30507-1G OR30507-5G
76574-334EA 17 USD
76574-334 76574-332
2-Indanol
2-Indanol
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 120…123 °C/12mmHg
Melting Pt: 67…72 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003800
CAS Number: 4254-29-9
EINECS: 224-230-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR