2-Bromo-4-methylanisole 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR3050-50G OR3050-250G OR3050-10G
76574-288EA 68 USD
76574-288 76574-290 76574-286
2-Bromo-4-methylanisole 98%
2-Bromo-4-methylanisole
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 134 °C/25mmHg
Melting Pt: 15.5 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00017783
CAS Number: 22002-45-5
EINECS: 244-705-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR