4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98% free radical

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl

OR30449-5G OR30449-25G OR30449-100G
76574-260EA 48 USD
76574-260 76574-264 76574-262
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 98% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Boiling Pt: 98 °C/3.75mmHg
Melting Pt: 33…39 °C
Density: 0.896 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR