Methyl-3-aminothiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3-amino-2-thiophenecarboxylate

OR30429-25G OR30429-100G OR30429-250G
76574-236EA 13 USD
76574-236 76574-238 76574-240
Methyl-3-aminothiophene-2-carboxylate
Methyl 3-aminothiophene-2-carboxylate
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Boiling Pt: 100…102 °C/0.1mmHg
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00009765
CAS Number: 22288-78-4
EINECS: 244-895-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR