6-Bromo-5-methoxy-2-pyridinecarbaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-Bromo-5-methoxypicolinaldehyde

OR303822-1G OR303822-500MG
76574-184EA 535 USD
76574-184 76574-182
6-Bromo-5-methoxy-2-pyridinecarbaldehyde
6-Bromo-5-methoxy-2-pyridinecarbaldehyde
Formula: C₇H₆BrNO₂
MW: 216.04 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD13193086
CAS Number: 329217-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR