(6-Morpholino-3-pyridinyl)-1-ethanone

Supplier: APOLLO SCIENTIFIC
Warning

OR303779-100MG OR303779-250MG
76574-176EA 80 USD
76574-176 76574-174
(6-Morpholino-3-pyridinyl)-1-ethanone
(6-Morpholino-3-pyridinyl)-1-ethanone
Formula: C₁₁H₁₄N₂O₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD09910348
CAS Number: 265107-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR