2-Chloro-6-methoxynicotinic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR303632-1G OR303632-250MG
76574-136EA 31 USD
76574-136 76574-138
2-Chloro-6-methoxynicotinic acid
2-Chloro-6-methoxynicotinic acid
Formula: C₇H₆NO₃Cl
MW: 187.5804 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD08236845
CAS Number: 503000-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR