Ethyl trimethylacetopyruvate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 5,5-dimethyl-2,4-dioxohexanoate

OR30152-1G
76573-874EA 13 USD
76573-874
Ethyl trimethylacetopyruvate
Ethyl trimethylacetopyruvate
Formula: C₁₀H₁₆O₄
MW: 200.24
Boiling Pt: 68…70 °C/0.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052319
CAS Number: 13395-36-3
EINECS: 236-478-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR