3-Amino-7-bromoisoquinoline

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 7-Bromoisoquinolin-3-amine

OR301335-100MG
76573-848EA 81 USD
76573-848
3-Amino-7-bromoisoquinoline
3-Amino-7-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040207
CAS Number: 1192815-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR