2,6-Dimethylbenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR30130-100G OR30130-25G OR30130-5G
76573-832EA 22 USD
76573-832 76573-834 76573-830
2,6-Dimethylbenzonitrile
2,6-Dimethylbenzonitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00037166
CAS Number: 6575-13-9
EINECS: 229-503-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR