2-Methylnicotinamide 97%

Supplier: APOLLO SCIENTIFIC

OR301074-1G
76573-676EA 82 USD
76573-676
2-Methylnicotinamide 97%
2-Methylnicotinamide
Formula: C₇H₈N₂O
MW: 136.1512 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909457
CAS Number: 58539-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR