1-Benzyl-3-pyrrolidinone

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-(Phenylmethyl)-3-pyrrolidinone1-Benzylpyrrolidin-3-one

OR301019-25G OR301019-5G
76573-626EA 13 USD
76573-626 76573-628
1-Benzyl-3-pyrrolidinone
1-Benzyl-3-pyrrolidinone
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 77…78 °C/0.01mmHg
Density: 1.091 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005342
CAS Number: 775-16-6
EINECS: 212-274-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR