5-Bromo-1-chloroisoquinoline

Supplier: APOLLO SCIENTIFIC
Warning

OR300068-250MG OR300068-1G
76573-136EA 52 USD
76573-136 76573-138
5-Bromo-1-chloroisoquinoline
5-Bromo-1-chloroisoquinoline
Formula: C₉H₅BrClN
MW: 242.4997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703278
CAS Number: 34551-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR