1-Amino-7-bromoisoquinoline 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Bromoisoquinolin-1-amine

OR300055-250MG OR300055-5G OR300055-1G
76573-118EA 121 USD
76573-118 76573-120 76573-122
1-Amino-7-bromoisoquinoline 97%
1-Amino-7-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374397
CAS Number: 215453-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR