3-Iodobenzylamine hydrochloride
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR2968-25G
OR2968-1G
OR2968-5G
76572-998EA
190
USD
76572-998
76572-996
76572-994
3-Iodobenzylamine hydrochloride
3-Iodobenzylamine hydrochloride
Formula:
C₇H₉ClIN MW: 269.5105 g/mol Melting Pt: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012857 CAS Number: 3718-88-5 EINECS: 223-068-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...