Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

OR29670-1G OR29670-250MG
76572-988EA 269 USD
76572-988 76572-986
Methyl-4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Methyl 4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Formula: C₈H₁₁BrN₂O₂
MW: 247.0891 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173775
CAS Number: 175276-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR