Methyl-3,5-dibromobenzoate 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3,5-dibromobenzoate

OR29245-5G OR29245-25G
76572-874EA 30 USD
76572-874 76572-872
Methyl-3,5-dibromobenzoate 98%
Methyl 3,5-dibromobenzoate
Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Melting Pt: 62…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00082648
CAS Number: 51329-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR